Would you like to log in again?
Microliter™ 700 Series Syringes with Luer Tip, Stoelting
Supplier: Stoelting
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Microliter™
51114 51117 51112 51116 51115 51113
95060-102EA 91.47 USD InStock
95060-102 95060-104 95060-098 95060-100 95060-106 95060-108
Microliter™ 700 Series Syringes with Luer Tip, Stoelting
Syringes Syringes for Chromatography
The 700 series syringe is the original hand-fitted Hamilton syringe.
This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL.
Ordering information: A variety of different terminations and needle options are available in this series.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty