Would you like to log in again?
Drosophila Diet, HiMedia
Supplier: HiMedia Laboratories
ID001-5L ID001-1L ID001-5L
95060-288EA 296.56 USD InStock
CA95060-288 CA95060-286 95060-288
Drosophila Diet, HiMedia
Cell Culture Supplements and Additives
Drosophila diet, ID001 is a modification of E
  • Modification of E. B. Lewis's original formula
  • Mimics the insect's normal food source
  • Provides the necessary nutrition for growth and development of fruit fly larvae
This medium mimics the insect's normal food source and provides the necessary nutrition for growth and development of fruit fly larvae.
B. Lewis's original formula for fruit fly larvae.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty