Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
NaCl 0.85% Medium, bioMerieux
Supplier: bioMérieux
20230 20070 20070 20230
95060-672PK 291.14 USD InStock
CA-95060-678 95060-672 CA-95060-672 95060-678
NaCl 0.85% Medium, bioMerieux
Microbiology Test Systems Microorganism Tests
Saline products are recommended for procedures that require the use of an isotonic diluent, as well as for use in preparing test suspensions of organisms.
  • Provides osmotic protection for microbial cells
  • Adjusts turbidity of bacterial suspensions
  • Maintains cell viability and integrity
Normal saline is used for preparing microbial suspensions when it is necessary to deliver a set number of microbes to an identification test battery, to antimicrobial agents, or to growth media used for disk susceptibility testing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty