Would you like to log in again?
API® 20 Microbial Identification Kits, bioMerieux
Supplier: bioMérieux
20701 20160 20050 20050 20600 20100 20300 20600 20160 20240 20100 20701 20300
95060-686PK 478.06 USD InStock
95060-686 95060-674 95060-688 95060-676 95060-682 95060-670 CA-95060-688 CA-95060-686 CA-95060-676 CA-95060-674 CA-95060-682 CA-95060-680 CA-95060-670
API® 20 Microbial Identification Kits, bioMerieux
Microbiology Test Systems Microorganism Tests
Standardized, miniature version of conventional methods conducting various microbial tests.
  • Standardized
  • Easy to use
  • Identify gram-negative, gram-positive, and anaerobic bacteria, yeasts, and other microorganisms
These microbial identification kits are used for identifying gram-negative, gram-positive, and anaerobic bacteria, yeasts, and other clinically significant microorganisms.
Additional reagents required see package insert for details
https://us.vwr.com/store/product/18630970/microbial-identification-reagents-biomerieux
API Web software required for Identification -purchase directly from bioMerieux.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty