Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VITEK® 2 Compact ID Cards, bioMerieux
Supplier: bioMérieux
21342 21347 21347 21346 21343 21346 21345 21341 21348 21343 21342 21345 21341
95060-696PK 553.7 USD InStock
95060-696 CA-95060-704 95060-698 CA-95060-702 97067-006 CA-95060-700 95060-694 95060-704 CA-95060-698 CA-95060-696 95060-700 CA-95060-694 95060-702
VITEK® 2 Compact ID Cards, bioMerieux
Microbiology Test Systems Microorganism Tests
Offers a comprehensive menu for the identification and antibiotic susceptibility testing of organisms.
  • Sealed against aerosols, splattering, spills, and other contaminants
  • Disposable waste reduced by over 80%
  • Size and shape of a playing card
VITEK® 2 card for Gram Negative organisms. The VITEK® 2 card is the size and shape of a playing card and contains 64 microwells.Each well contains identification substrates or antimicrobials. VITEK® 2 offers a comprehensive menu for the identification and antibiotic susceptibility testing of organisms.The VITEK® 2 test card is sealed, which minimizes aerosols, splattering, spills, and personal contamination. Disposable waste is reduced by more than 80% over other methods.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty