Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
API® ID 32 Microbial Identification Kits, Biomerieux
Supplier: bioMérieux
32200 32300 32700 32600 32500 32700 32200 32600 32300 32500 10300
95060-710PK 643.34 USD InStock
CA-95060-716 95060-710 CA-95060-714 95060-666 CA-95060-712 CA-95060-710 95060-716 95060-718 95060-712 CA-95060-718 95060-714
API® ID 32 Microbial Identification Kits, Biomerieux
Microbiology Test Systems Microorganism Tests
Standardized, miniature version of conventional methods conducting various microbial tests
  • Standardized
  • Easy to use
  • Identify gram-negative, gram-positive, and anaerobic bacteria, yeasts, and other microorganisms
These microbial identification kits are used for identifying gram-negative, gram-positive, and anaerobic bacteria, yeasts, and other clinically significant microorganisms.
ID 32/Rapid ID 32 kits are designed for the identification of Enterobacteriaceae, Anerobes, Staph, Strep, and Yeast species.
Additional reagents required see package insert for details
https://us.vwr.com/store/product/18630970/microbial-identification-reagents-biomerieux
API Web software required for Identification -purchase directly from bioMerieux.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty