Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Irradiated 100 mm Plates Media, BioMerieux
Supplier: bioMérieux
M1060-TI M1025-TI 43169 43711 M1219-IR 47071 M1060-IR M1253-TI M1025-IR M1049-IR 43287 M1040-IR M1040-TI
95061-298PK 84.82 USD InStock
95061-298 95061-330 95061-324 95061-334 89131-936 95061-410 95061-398 95061-314 95061-318 95060-814 95061-294 95060-810 95060-832
Irradiated 100 mm Plates Media, BioMerieux
Media for Microbiology Prepared Media for Microbiology
These contact plates can be used for surface environmental monitoring.
Sabouraud Dextrose Agar is used for the detection and enumeration of viable fungal contaminants present on surfaces and equipment of sanitary importance.
Irradiated Tryptic Soy Agar (TSA) is used for the quantitation of viable microbial contaminants from surfaces and equipment in controlled environments also used for viable air monitoring.
Irradiated Dey-Engley Neutralizing Agar is used for the detection and enumeration of micro-organisms that are present in environments that contain antimicrobial chemical, such as surfaces that are of sanitary importance.
Irradiated Tryptic Soy Agar (TSA) with Lecithin and Polysorbate 80 is used for the quantitation of viable microbial contaminants from surfaces and equipment in controlled environments.
Trypcase soy 3P™ agar with neutralizers and lactamase is intended for the monitoring of microbial contamination in pharmaceutical industry cleanrooms.
Ordering information: Made to order items have a lead time of 42 days.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty