Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Thioglycollate Broth with Resazurin, Biomerieux
Supplier: bioMérieux
44001 44001
95060-842CS 146.21 USD InStock
95060-842 CA95060-842
Thioglycollate Broth with Resazurin, Biomerieux
Media for Microbiology Prepared Media for Microbiology
Mostly used for detection anaerobic bacteria.
  • Detects anaerobic bacteria
  • Also allows detection of aerobic bacteria
Mostly used for detection of anaerobic bacteria but also allows detection of aerobic bacteria recommended by USP/EP/JP for sterility testing of pharmaceutical articles.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty