Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sterility Culture Media Range, BioMerieux
Supplier: bioMérieux
44011 44021 44021 42634 42633 44011
95060-796CS 76.98 USD InStock
95060-796 95060-794 CA95060-846 95060-844 CA95060-844 95060-846
Sterility Culture Media Range, BioMerieux
Media for Microbiology Dehydrated Media for Microbiology
General purpose medium for cultivation of a wide variety of microorganisms recommended by USP/EP/JP for sterility testing of pharmaceutical articles.
  • For both fastidious and non-fastidious microorganisms
  • Accomodates a wide variety
  • Effective
A general-purpose medium used for the cultivation of fastidious and non-fastidious microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty