Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Culture Media Range for Media Fill Test, bioMerieux
Supplier: bioMérieux
51102 51103 51101 51104
95060-860EA 203.33 USD InStock
95060-860 95060-858 95060-854 95060-856
Culture Media Range for Media Fill Test, bioMerieux
Media for Microbiology Dehydrated Media for Microbiology
The 3P™ Culture Media Range for Media Fill Test uses unique color indicators in media fill tests.
  • Unique color indicator
  • Optimized for quick aerobic and anaerobic contamination detection
  • Greater ease of use
  • Fewer false positive and false negative results
Bringing a new dimension to the media fill test with a unique color indicator.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty