Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
BioBall®, MultiShot 550, bioMerieux
Supplier: bioMérieux
416600 56008 56012 56014 56003 56002 56021 56019 56007 56001 56013 56009 56016 56016 56018 56005 56018 56011 56015 56017 56012 56013 56015 56004 56021 56017 56006 56014
10500-244PK 1000.21 USD InStock
10500-244 10500-242 10500-240 CA95060-872 95060-864 CA95060-870 95060-880 95060-882 CA95060-878 CA95060-876 95060-866 95060-868 CA95060-874 10500-238 10500-236 10500-234 10500-232 10500-230 75997-536 95060-872 CA95060-882 95060-874 95060-870 CA95060-868 CA95060-866 95060-876 10500-228 95060-878
BioBall®, MultiShot 550, bioMerieux
Microbiology Test Systems Microorganism Tests
BioBall® MultiShot 550 is ideal for testing the efficacy of antimicrobial preservation.
  • Unprecedented accuracy for quantitative microbiological quality control
  • BioBall® is an accredited reference material under ISO Guide 34 standard
  • Easy to use-no preparation or pre-incbation
Offers unparalleled precision, ease of use and is ideal for industrial applications requiring a known high level of inoculum.
BioBall® contains a precise number of microorganisms in a water soluble ball delivering unprecedented precision and accuracy for Quantitative Microbiological Quality Control.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty