Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
BioBall® SingleShot 30, BioMerieux
Supplier: bioMérieux
56028 56029 56022 56044 56033 56045 56040 56034 56038 56041 56092 56042 56032 56030 56039 56035 56027 56029 56026 56033 56031 56040 56023 56045 56035 56039 56028 56034 56032 56024 56026 56031 56042 56024 56038 56044 56023 56043 56022 412690 56046 56030 56041
10500-300PK 668.95 USD InStock
CA95060-908 10500-300 95060-918 CA95060-906 CA95060-904 CA95060-894 95060-884 CA95060-892 95060-886 CA95060-890 CA95060-914 95060-902 97067-096 CA95060-912 97067-094 95060-904 95060-888 CA95060-898 95060-920 CA95060-910 CA95060-896 95060-900 97067-098 95060-906 CA95060-918 CA95060-916 95060-908 10500-294 CA95060-884 95060-894 95060-896 95060-890 95060-892 CA95060-902 95060-914 95060-916 CA95060-900 CA95060-920 95060-910 95060-898 CA95060-888 95060-912 CA95060-886
BioBall® SingleShot 30, BioMerieux
Microbiology Test Systems Microorganism Tests
BioBall® contains a precise number of microorganisms in a water soluble ball designed for a single inoculation
  • Precise number of microorganisms
  • Unprecedented accuracy
  • Water-soluble delivery system
Offers unparalleled precision, ease of use & is ideal for industrial applications requiring a known high level of inoculum. SingleShot offers a mean between 28cfu and 33cfu, with a standard deviation of 3cfu or less unless otherwise stated.
Ideal positive control for media QC, and a positive control for pathogen testing in the food industry
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty