Would you like to log in again?
BioBall®, High Dose 10KX10, bioMerieux
Supplier: bioMérieux
56055 413831 56051 56144 56160 56054 56159 56143 413779 413777 56052 56158 56050 413776 56053 56049 413778
10500-214PK 387.14 USD InStock
10500-214 10500-226 10500-222 95060-928 10500-220 95060-930 95060-936 95060-924 95060-938 95060-926 95060-932 10500-218 10498-672 95060-934 10498-674 10500-216 95060-922
BioBall®, High Dose 10KX10, bioMerieux
Microbiology Test Systems Microorganism Tests
Designed for a single inoculation at a high cfu level
  • Unprecedented accuracy for quantitative microbiological quality control
  • Easy to use-no preparation or pre-incubation
  • BioBall® is an accredited reference material under ISO Guide 34 standard
BioBall® contains a precise number of microorganisms in a water soluble ball delivering unprecedented precision and accuracy for Quantitative Microbiological Quality Control. HighDose 10K; mean of between 8,000 and 12,000 cfu with a standard deviation of ≤ 15%.
Ideal for positive control testing where a higher cfu is required.
Certifications: Strains are USP compliant.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty