Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
BioBall® MultiShot 10E8, bioMerieux
Supplier: bioMérieux
56145 56147 56148 410106 56146
97067-104PK 1335.18 USD InStock
97067-104 97067-244 95060-940 95060-942 95060-944
BioBall® MultiShot 10E8, bioMerieux
Microbiology Test Systems Microorganism Tests
BioBall® MultiShot 10E8 is ideal for testing the efficacy of antimicrobial preservation.
  • Unprecedented accuracy for quantitative microbiological quality control
  • Easy to use-no preparation or pre-incubation
  • BioBall® is an accredited reference material under ISO Guide 34 standard.
BioBall® contains a precise number of microorganisms in a water soluble ball delivering unprecedented precision and accuracy for Quantitative Microbiological Quality Control. Pharmaceutical and cosmetic preparations require antimicrobial activity to prevent microbial proliferation or to limit the effects of contamination. BioBall® MultiShot 10E8 is is ideal for this application.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty