Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
McFarland Standards Kit, Biomerieux
Supplier: bioMérieux
McFarland
70900 70900
95060-984EA 181.34 USD InStock
95060-984 CA-95060-984
McFarland Standards Kit, Biomerieux
Control reagents
McFarland standards are used as a reference for adjusting the turbidity of bacterial suspensions so that the number of bacteria will be within a given range to standardize microbial testing.
  • Stable suspensions
  • Allows for visual comparison with bacterial suspensions
Standards are economical and easy to use, with straightforward comparison with bacterial suspensions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty