Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Non Irradiated Contact Plates Media, BioMerieux
Supplier: bioMérieux
C6047 C6064 C6045
95061-200PK 40.3 USD InStock
95061-200 95061-180 95061-172
Non Irradiated Contact Plates Media, BioMerieux
Media for Microbiology Prepared Media for Microbiology
For detection and enumeration of microorganisms present on surfaces of sanitary importance.
Tryptic Soy Agar with Lecithin and Polysorbate 80 is used for the quantitation of viable microbial contaminants from surfaces and equipment in controlled environments.
D/E agar is a medium used for the detection and enumeration of microorganisms that are present in environments that contain antimicrobial chemicals, such as surfaces that are of sanitary importance.
Sabouraud Dextrose Agar w/Lecithin and Polysorbate 80 is used for the detection and enumeration of viable fungal contaminants present on surfaces and equipment of sanitary importance.
Ordering information: Made to order Items have a lead time of 42 days.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty