Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Irradiated Contact Plates Media, BioMerieux
Supplier: bioMérieux
412145 46991 C6045-IR 43691 43699 C6045-IX C6064-TI C6064-IR 410134 C6047-TI 412205 410183 C6045-TI
95061-176PK 61.53 USD InStock
95061-176 89131-944 95061-186 95061-174 95061-202 95061-178 95061-206 89347-982 89131-890 89368-796 95060-830 97067-250 95060-828
Irradiated Contact Plates Media, BioMerieux
Media for Microbiology Prepared Media for Microbiology
These contact plates can be used for surface environmental monitoring.
Irradiated Count-Tact 3P agar is used for the monitoring of microbial contamination in industrial and hospital clean room environments.
Dey-Engley agar is a medium used for the detection and enumeration of micro-organisms that are present in environments that contain antimicrobial chemicals, such as surfaces that are of sanitary importance.
Irradiated Tryptic Soy Agar (TSA) with Lecithin and Polysorbate 80 is used for the quantitation of viable microbial contaminants from surfaces and equipment in controlled environments.
Sabouraud Dextrose Agar with Lecithin and Polysorbate 80 is for isolation and cultivation of fungi from surfaces sanitized with quaternary ammonium compounds.
Ordering information: Made to order items have a lead time of 42 days.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty