Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Irradiated 150 mm Plates Media, BioMerieux
Supplier: bioMérieux
L2037-IR L2037-TI
95061-262PK 104.2 USD InStock
95061-262 95061-260
Irradiated 150 mm Plates Media, BioMerieux
Media for Microbiology Prepared Media for Microbiology
These contact plates can be used for surface environmental monitoring.
Tryptic Soy Agar is a general-purpose solid medium used for the cultivation and enumeration of a wide variety of fastidious and non-fastidious micro-organisms. Also known as Soybean-Casein Digest Agar (USP).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty