Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Non Irradiated Media 100 mm Plates, BioMerieux
Supplier: bioMérieux
M1006 410190 M1060 M1031 43596 M1025 43031 M1042 M1017 M1027 M1040 M1067 M1070 A0101 46979 M1065 M1038
95061-320PK 64.45 USD InStock
95061-320 95061-274 95061-284 95061-346 95061-158 95061-300 95061-344 95061-310 95061-328 95061-338 95061-304 95061-308 97067-048 97067-258 95060-806 95060-822 95061-292
Non Irradiated Media 100 mm Plates, BioMerieux
Media for Microbiology Prepared Media for Microbiology
General purpose medium for the cultivation of a wide variety of microorganisms.
Anaerobic Blood Agar (CDC Formulation) is used for the primary isolation and cultivation of anaerobic micro organism.
Baird Parker Agar is used for the selective isolation and enumeration of coagulase-positive staphylococci and may also be used for identification of staphylococci on the basis of lecithinase activity.
Cetrimide Agar is a selective and differential medium used in the presumptive identification and isolation of pseudomonas aeruginosa.
Legionella BCYE medium without L-cysteine is used to confirm or exclude the presence of Legionella in water by subculture of typical colonies obtained previously using Legionella GVPC medium. This medium should be inoculated at the same time as the BCYE medium with L-cysteine. If the strain does not grow on BCYE medium without L-cysteine, it can be confirmed as belonging to the genus Legionella.
Legionella GVPC agar has been specially developed for the enumeration of legionella in water.
M Endo Les Agar is a medium used for the enumeration of coliforms in water by the membrane filtration technique as recommended by the Standard Methods for the Examination of Water and Waste water.
Potato Dextrose Agar is recommended for the identification, cultivation and enumeration of yeasts and molds and to induce sporulation in all fungi. It has been used for cultivation and isolation of molds and yeasts in dairy and various food products as recommended by the American Public Health Association.
Sabouraud Dextrose Agar is used for the detection and enumeration of viable fungal contaminants present on surfaces and equipment of sanitary importance. Sabouraud Dextrose Chloramphenicol Agar (SDC) is used for the cultivation of dermatophytes and both pathogenic and non-pathogenic yeasts and molds.
Tryptic soy agar TSA is used for the cultivation and enumeration of a wide variety of fastidous and non-fastidous micro-organisms. Tryptic soy agar with 5% sheep blood (Blood Agar) is a general purpose medium used for cultivation enumeration and hemolysis determination of fastidious and non-fastidious micro-organism.
Tryptic soy agar with 5% Sheep Blood and 0.1% Esculin is a non-selective, differential medium for the cultivation, enumeration and differentiation of organisms based on hemolysis and esculin hydrolysis reactions.XLD Agar is a selective differential medium for the isolation of gramnegative enteric bacteria.
Solid medium used for the detection and enumeration of aerobic mesophilic bacteria, yeasts and molds according to ISO 21149 standard for cosmetic products. Standard Methods Agar is a media used for enumeration of viable micro-organisms in water, waste water, foods and dairy products.
Ordering information: Made to order items have a lead time of 42 days.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty