Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Thioglycollate Broth with Resazurin Tubed Culture Media, Biomerieux
Supplier: bioMérieux
P1700 P1280-SC
95061-454CS 232.6 USD InStock
95061-454 95061-449
Thioglycollate Broth with Resazurin Tubed Culture Media, Biomerieux
Media for Microbiology Prepared Media for Microbiology
Fluid Thioglycolate Medium (FTM) is used for the cultivation of aerobic, anaerobic and microaerophilic organisms.
  • For detection of anaerobic bacteria
  • Multiple sizes available
Mostly used for detection of anaerobic bacteria but also allows detection of aerobic bacteria recommended by USP/EP/JP for sterility testing of pharmaceutical article. FTM and FTM w/Polysorbate 80 are used as enrichment media for sterility testing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty