Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sabouraud Dextrose Agar, bioMerieux
Supplier: bioMérieux
P2070
95061-470CS 475.77 USD InStock
95061-470
Sabouraud Dextrose Agar, bioMerieux
Media for Microbiology Prepared Media for Microbiology
Sabouraud Dextrose Agar is used for the detection and enumeration of viable fungal contaminants present on surfaces and equipment of sanitary importance.
  • Media for the isolation and cultivation of fungi
  • 10 x 500 mL
Upon receipt, store plates at 2 – 8 °C away from direct light in an inverted position. Store bottles according to the product label in an upright position. Store plates in original outer wrapping until ready to use. Media can be inoculated up to the expiration date and incubated for the appropriate incubation period.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty