Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Thioglycollate Broth with Resazurin, Biomerieux
Supplier: bioMérieux
P2770-RS P2360
95061-492CS 261.98 USD InStock
95061-492 95061-506
Thioglycollate Broth with Resazurin, Biomerieux
Media for Microbiology Prepared Media for Microbiology
Fluid Thioglycollate Medium (FTM) is used for the cultivation of aerobic, anaerobic and microaerophilic organisms
  • For the cultivation of aerobic, anaerobic and microaerophilic organisms
  • Multiple sizes available
Mostly used for detection of anaerobic bacteria but also allows detection of aerobic bacteria recommended by USP/EP/JP for sterility testing of pharmaceutical article. FTM as described by Brewer was formulated for the rapid growth of anaerobes and aerobes. FTM is used for sterility testing and for the cultivation of anaerobes, aerobes and microaerophiles. The FDA Bacteriological Analytical Manual, the Official Methods of Analysis of AOAC International and Compendium of Methods for the Microbiological Examination of Foods recommend FTM for the examination of food and for determining the phenol coefficient and sporicidal effects of disinfectants.
FTM and FTM w/Polysorbate 80 are used as enrichment media for sterility testing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty