Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Tryptic Soy Agar Bottles, Biomerieux
Supplier: bioMérieux
P2050 P2050
95061-510CS 319.75 USD InStock
95061-510 CA-95061-510
Tryptic Soy Agar Bottles, Biomerieux
Media for Microbiology Prepared Media for Microbiology
Tryptic Soy Agar is a general-purpose solid medium used for the cultivation and enumeration of a wide variety of fastidious and non-fastidious microorganisms.
  • General purpose medium
  • 10 x 500 mL
Store according to the package label in an upright position in the dark. Do not freeze. Allow warming of medium to room temperature before inoculation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty