Would you like to log in again?
Sabouraud Dextrose Agar Tubed Culture Media, bioMerieux
Supplier: bioMérieux
T8008
95061-572CS 97.9 USD InStock
95061-572
Sabouraud Dextrose Agar Tubed Culture Media, bioMerieux
Media for Microbiology Prepared Media for Microbiology
Sabouraud Dextrose Agar is used for the detection and enumeration of viable fungal contaminants present on surfaces and equipment of sanitary importance.
  • For detection of viable fungal contaminants
  • 20 x 20 mL
Media for the isolation and cultivation of fungi. Sabouraud Dextrose Agar contains enzymatic digests of casein and animal tissue that are excellent sources of nitrogen, amino acids, and peptides. Dextrose is the carbohydrate source. Agar is the solidifying agent.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty