Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
HBIO Petri Dish Platinum Electrodes for Tissue Slices Chamber Kita, BTX
Supplier: BTX
45-0505 45-0500 45-0510 45-0507 45-0509
95062-350EA 4506.63 USD InStock
95062-350 95062-372 95062-360 95062-364 95062-370
HBIO Petri Dish Platinum Electrodes for Tissue Slices Chamber Kita, BTX
Electrodes pH Electrodes
Electrodes and chambers for delicate and/or difficult tissue transfection.
  • Simple and easy to use
  • Compatible with BTX Systems such as Gemini X2 and ECM 2001+
  • Comprised of two parts, glass petri dish and electrode wand
  • Wand, cables included
The Petri dish platinum electrodes are designed for delicate and/or difficult ex vivo tissue transfection. Ex vivo electroporation is an efficient, effective method to introduce genes, drugs or any number of molecules into ex-plant tissues. A common application is mouse brain slice for studying neuronal development. This specialty electrode makes transfection quick and simple and is compatible with the BTX ECM 830, ECM 2001+, Gemini X2 and Legacy ECM 2001 generators.
The electrode is comprised of two parts, the glass petri dish electrode and electrode wand. The glass petri dish contains a square shaped platinum electrode chamber to secure the tissue. The wand incorporates an identical shaped platinum electrode, which is placed over the chamber to complete electroporation. This sandwich configuration ensures a homogeneous field of energy for optimum transfection.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty