Would you like to log in again?
Single-Edge Razor Blade Pop-Up Dispenser, Accutec Blades
Supplier: AccuTec Blades
62-0325-0000
95062-667CS 1517.91 USD InStock
95062-667
Single-Edge Razor Blade Pop-Up Dispenser, Accutec Blades
Razors and Blades
The Pop-up dispenser enables the user to access a blade without having to touch it before use.
Simple operation allows the blade to be unwrapped and made accesible with the push of a button. This dispenser contains 100 Gem 3 Facet carbon steel single-edge blades. Three facet blades provide an extra-keen edge that enables intricate cutting.
Ordering information: Contains 100 GEM 3 facet single edge blades with an aluminum back.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty