Would you like to log in again?
NEFA Linearity Set Solution, Wako
Supplier: Wako
276-76491 997-76491 276-76491 993-35191 995-34791 997-76491 999-34691 991-34891
97000-016EA 211.23 USD InStock
97000-016 97000-014 CA11001-412 10753-190 97000-012 CA10753-190 97000-010 80076-166
NEFA Linearity Set Solution, Wako
Standards Color Reference and Spectroscopy Standards
The NEFA Linearity Set is a 2.5 mEq/L solution used to verify the measurable range of the NEFA HR(2) assay.
This product is only used with Wako’s NEFA HR(2) series of reagents.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty