Would you like to log in again?
Slimsette™ M511 Tissue Processing/Embedding Cassettes, Simport
Supplier: Simport
SLIMSETTE™
M511-9 M511-16 M511-12 M511-18 M511-8 M511-4 M511-16 M511-18 M511-7 M511-6 M511-12 M511-10 M511-6 M511-8 M511-11 M511-17 M511-17 M511-2 M511-5 M511-2 M511-10 M511-4 M511-5 M511-11 M511-3 M511-9 M511-3 M511-7
97000-418CS 567.75 USD InStock
97000-418 CA97000-428 97000-424 CA97000-432 97000-444 97000-422 CA97000-430 97000-428 CA97000-436 CA97000-434 97000-426 97000-442 97000-420 97000-440 CA97000-418 CA97000-438 CA97000-444 97000-434 CA97000-422 97000-432 CA97000-442 CA97000-420 97000-438 CA97000-426 97000-436 CA97000-424 CA97000-440 97000-430
Slimsette™ M511 Tissue Processing/Embedding Cassettes, Simport
Embedding Cassettes Tissue Processing Cassettes
These compact cassettes feature a unique recessed cover that allows more cassettes to be stacked in automatic labeling machines and in storage cabinets
Cassettes are molded from acetal polymer and are completely resistant to the chemical action of histological solvents. Four 13mm square compartments have openings in the cover and base for maximizing fluid exchange and ensuring proper drainage. The one-piece integral lid eliminates the need for separate steel lids and can be opened and closed as often as necessary. Lids relock securely without danger of specimen loss. Cassettes feature large labeling areas for easy identification. The anterior writing area is slanted at a 45° angle for suitability with certain types of cassette labeling instruments. Cassette colors are non-cytotoxic and non-metallic.
Ordering information: Each case contains three dispenser boxes of 500 cassettes with the lids closed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty