Would you like to log in again?
Germicidal Workstation, Plas-Labs™
Supplier: Plas-Labs
820-TCH/EXP 820-TCH
97000-990EA 6433.6 USD InStock
CA97000-990 97000-990
Germicidal Workstation, Plas-Labs™
Glove Boxes and Glove Bags
Ideal for animal necropsy, tissue culture processing, and PCR work
Workstation can be used as a safety enclosure, teaching demonstration hood, toxicology or pathology workstation, or still air hood. The main housing unit is formed from one piece of 1cm (3/8") thick optically clear acrylic for total visibility. Formed plastic bottom has a spill guard. Workstation contains both fluorescent and UV germicidal lights with a safety switch to prevent operating both lights at the same time. Other features include electrical overload protection, illuminated controls, removable top cover for easy access to lightbulbs, and ground key cock valve for gas circulation. The sash opening is 69.9Wx21.6Hcm (271/2x81/2").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty