Would you like to log in again?
Compact Glove Boxes, Plas-Labs™
Supplier: Plas-Labs
830CP-30/EXP 830-ABC/EXP 830CP-21/EXP 830-CP-10 830-CP-21 830CP-10/EXP 830CP-11/EXP 830-ABC 830CP-20/EXP 830-ABB/EXP 830CP-31/EXP 830-CP-20 830-ABD 830-ABB 830-CP-11 830-CP-31 830-CP-30 830-ABD/EXP
97001-002EA 14626.27 USD InStock
97001-002 CA97001-000 97001-004 CA97001-002 CA97001-004 97001-000 CA97000-994 97000-996 97000-994 32940-008 CA97000-998 10718-112 CA32940-008 CA97000-996 CA10718-112 10718-114 97000-998 CA10718-114
Compact Glove Boxes, Plas-Labs™
Glove Boxes and Glove Bags
Designed specifically for use when weighing hazardous materials that must be contained, glove boxes provide cleanliness, safety, portability, and a draft-free atmosphere for the maximum effectiveness of your balance
Top section is removable for easy installation of equipment, such as analytical balances, top loaders, bench scales, and moisture analyzers. Basic features include four purging gas valves (two on the transfer chamber and two on the main chamber), a white leveling tray for easy transferring of liquids, white ambidextrous Hypalon® gloves for superior chemical and UV resistance, a pressure relief valve that minimizes glove "fight back," and replaceable HEPA and organic vapor canister filters. All clamps are stainless steel and completely adjustable. Transfer chamber inner dimensions: 30.5Lx27.9Dcm (12x11").
Hospital-grade multiple electrical outlet strip is UL listed and CSA certified. CE marked.
Beneficial when working with toxic substances, asbestos fibers, sewage residue, and harmful liquid vapors. Ideal for determining the dry weight or moisture content of aqueous solutions, adhesives, cereals, toxic chemicals, pharmaceuticals, paper, plastics, and light radioactive materials. Minimize product degradation caused by moisture absorption.
Ordering information: Supplied with one-year manufacturer's warranty and instruction manual. Custom sizes are available. Models with additional glove ports are available. Exterior dimensions include transfer chamber. Allow 10cm (4") for elbow valve opposite transfer chamber. Contact your Avantor sales representative for information.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty