Would you like to log in again?
Cabinet Accessories, Justrite®
Supplier: Justrite
29937 29961 29945 29938 25950 29965 29940 25933 29936 29930 29980 29939 29934 24168 29017 29962 29951 29972 29907 29968 29973 29950 29953 29960 24115 29018 29949 24167 29958 29935 29942 29948
10836-764EA 254.44 USD InStock
10836-764 97001-322 10836-886 10836-788 10836-888 10836-882 97001-326 10836-884 97001-328 97001-320 97001-318 10836-890 10836-770 10836-874 97001-310 10836-776 10836-876 97001-312 10836-778 97001-336 97001-314 10836-772 97001-316 10836-872 97001-338 10836-774 10836-878 97001-330 82011-464 97001-308 82011-462 10836-880
Cabinet Accessories, Justrite®
Furniture Shelves
Cabinet accessories offer an opportunity to expand storage and add new convenience to existing cabinets, or customize new cabinets with additional shelves to meet specific workflow needs.
  • Expand storage
  • Add new convenience
  • Customize new cabinets
Polyethylene trays offer additional protection and have a slightly raised curved edge to contain spills
SpillSlope™ galvanized steel shelves hold up to 159 kg (350 lbs.) and adjust on 5.7 cm (21/4") increments for versatile storage.
They may also be used in the bottom sill area to catch incidental spills and leaks.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty