Would you like to log in again?
InLab® Expert Pro 3-in-1 pH Electrodes, Double Junction, METTLER TOLEDO®
Supplier: Mettler-Toledo
InLab®
51343104 51343103 51343101 51343104 51343103 51343105 51343101 51343102 51343105 51343102
97001-610EA 429.87 USD InStock
97001-610 CA97001-610 97001-612 CA97001-612 97001-614 CA97001-614 97001-616 CA97001-616 97001-618 CA97001-618
InLab® Expert Pro 3-in-1 pH Electrodes, Double Junction, METTLER TOLEDO®
Electrodes pH Electrodes
Electrodes are ideal for a wide range of applications, including emulsions, suspensions, effluent water, and solutions containing proteins and sulphides.
  • Integrated temperature probe
  • Solid polymeric electrolyte
  • No junction required
  • ARGENTHAL™ lead-off and silver ion trap
  • PEEK™ shaft
  • High chemical resistance
They feature an integrated temperature probe, a solid polymeric electrolyte that requires no junction; an ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures; and a robust high-grade PEEK™ shaft for high chemical resistance. Electrodes with MultiPin™ connectors feature a low-resistance, universal glass membrane. Diameter: 12 mm. Lead length: 1.2 m, except for 97001-618 which is 2 m.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty