Would you like to log in again?
InLab® Viscous Combination pH Electrode, METTLER TOLEDO®
Supplier: Mettler-Toledo
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
InLab®
51343150 51343150
97001-628EA 840 USD InStock
97001-628 CA97001-628
InLab® Viscous Combination pH Electrode, METTLER TOLEDO®
Electrodes pH Electrodes
<bold>Electrode is designed for viscous and highly glutinous samples
  • SteadyForce™ reference system
  • S7 screw head
It features a SteadyForce™ reference system with under-over pressure to guarantee electrode discharge and prevent contamination. S7 screw head connects with different cables, so electrode may be used with a wide range of meters.
</bold>
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty