Would you like to log in again?
InLab® Micro Combination pH Electrode, METTLER TOLEDO®
Supplier: Mettler-Toledo
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
InLab®
51343160 51343160
97001-642EA 467 USD InStock
97001-642 CA97001-642
InLab® Micro Combination pH Electrode, METTLER TOLEDO®
Electrodes pH Electrodes
Electrode is designed for measuring in very small vessels, such as vials or cuvettes
  • ARGENTHAL™ lead-off and silver ion trap
  • Safelock™ ABS closing ring
  • S7 screw head connector
Features ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures. Safelock™ ABS closing ring as reference junction makes refilling more accessible and ensures complete sealing when closed. S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty