Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Dual-Rack Garment Racks, Stainless Steel, Bandy
Supplier: Bandy
GRK-DSL-48 GRK-DSL-72 GRK-DSL-84 GRK-DS-48 GRK60005 GRK-DSL-60 GRK-DS-84 GRK-DS-60 GRK-DS-72
97002-020EA 5866.73 USD InStock
97002-020 97002-018 98000-592 97002-016 98000-590 97002-014 98000-596 24450-006 98000-594
Dual-Rack Garment Racks, Stainless Steel, Bandy
Furniture Gowning Racks
Garment racks feature a 2.5x7.6cm (1x3"), 16-gauge, Type 304 stainless steel tube frame with an electropolished or No
4 finish. Available with a detachable or non-detachable hanger format. Hanger positions at 7.6cm (3") intervals on both sides. Racks include adjustable leveling feet.
Ordering information: Racks with detachable hanger format do not include hangers.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty