Would you like to log in again?
Heat-On Blocks, Radleys/Brinkmann®, Heidolph
Supplier: Heidolph
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
015882070CA 15852010 15882230 15882110 15882050 15882180 015882120CA 15882260 015882300CA 015882220CA 15882040 015882230CA 015882130CA 015882290CA 015882310CA 015852020CA 15882090 015882010CA 15882010 15882280 15882080 15882290 015882250CA 15882190 015852030CA 015882280CA 015882020CA 015882260CA 015882180CA 015882160CA 015882030CA 015882210CA 15882210 15882130 15852030 015882056 015882040CA 15852000 015882050CA 15852020 15882030 015882110CA 15882120 15882270 15882170 015882240CA 015882000CA 15882070 15882000 15882250 15882160 15882060 015882060CA 015882170CA 15882200 15882310 015852010CA 015882190CA 015852000CA 015882200CA 015882100CA 015882080CA 15882100 15882020 15882220 15882300 15882240 015882270CA
97002-076EA 526.63 USD InStock
CA11009-010 CA11008-890 CA11008-970 97002-076 97002-032 CA11008-884 CA11008-960 97002-030 CA11008-886 97002-074 CA11008-962 CA11008-888 97002-072 CA11008-964 97002-070 CA11009-008 CA11008-968 97002-038 CA11009-006 97002-036 CA11009-004 97002-034 97002-078 CA11009-002 CA11009-000 97002-080 CA11008-994 CA11008-950 97002-088 97002-044 CA11008-952 CA11008-996 97002-086 97002-042 CA11008-998 CA11008-954 97002-040 97002-084 CA11008-956 97002-082 CA11008-958 97002-048 97002-046 97002-090 CA11008-990 CA11008-992 97002-098 97002-054 CA11008-982 97002-096 97002-052 CA11008-984 97002-094 97002-050 CA11008-986 97002-092 CA11008-988 CA11008-948 97002-056 10145-578 CA11008-980 CA11008-972 97002-066 CA11008-974 97002-064 97002-062 97002-100 97002-068
Heat-On Blocks, Radleys/Brinkmann®, Heidolph
Block Heaters Blocks
A safer, cleaner alternative to oil baths and heating mantles, blocks are for heating and stirring one to two round bottom flasks at a time, up to 200°C (392°F) or 260°C (500°F) 20 minutes or less
Lightweight, solid aluminum blocks heat evenly and feature a deep well design for rapid heat transfer, so flasks will not stick or crack. Polymer or anodized coating provides strong chemical resistance. Blocks have two probe holes for temperature probe (not included), and fit on all common hotplates up to 145mm in diameter.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty