Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Blood Drawing Chairs, Adjustable Height, Med-Care Manufacturing

Supplier: Ward's
12CPABLUE 12CPADOVE
97002-115EA 3035.18 USD InStock
97002-115 97002-114
Blood Drawing Chairs, Adjustable Height, Med-Care Manufacturing
Chairs
Comfortable, molded plastic chairs feature hydraulically adjustable height and two adjustable padded side arms.

  • A third, flip-up padded arm provides safety and comfort for patient and technician
  • Welded steel construction
  • Durable plastic seat is easy to clean

Adjustable seat: 50.8 to 68.6 cm (20 to 27")
Side arms are 17.8 cm (7") wide
Side arm height adjustment 17.8cm (7")

Ordering information: Five-year manufacturer's warranty on chair frame. One-year manufacturer's warranty on vinyl upholstery. Contact your VWR sales representative for additional color choices.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty