Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Flexible Vent Assemblies, Scientific Plastics
Supplier: Scientific Plastics
F160L164F150 F160L078F-150 F2738565150 F2738567-150 F160L06XF150
97002-242EA 550.24 USD InStock
97002-242 97002-240 97002-256 97002-254 97002-244
Flexible Vent Assemblies, Scientific Plastics
Furniture Casework
Venting systems are designed for use in acid and solvent cabinets.
  • All vents are manufactured from acid-resistant polyolefin material
  • Non-squeezed top
The flexible tubing design allows systems to be installed around potential obstacles within existing cabinet structures. A 3 m (10') length of accessory tubing is available separately for field modifications.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty