Would you like to log in again?
LE501 Redox Combination Electrode, METTLER TOLEDO®
Supplier: Mettler-Toledo
51340338 51340338
97002-310EA 237 USD InStock
CA97002-310 97002-310
LE501 Redox Combination Electrode, METTLER TOLEDO®
Electrodes Redox Electrodes
Refillable electrode is for use with METTLER TOLEDO® pH meters in laboratory water analysis.
  • Refillable
  • BNC connector
Features BNC connector.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty