Would you like to log in again?
Autoclavable Carts and Accessories, Metro™
Supplier: Metro International
LC2S2436 LC2436AC LC2S2436 LC2S1824 LC3S1824 LC1824AC LC3S2436 LC3S2436 LC2436AC LC1824AC LC2S1824 LC3S1824
97002-848EA 990.48 USD InStock
CA97002-848 CA97002-846 97002-848 CA97002-844 97002-846 CA97002-842 97002-844 CA97002-840 CA97002-850 97002-842 97002-850 97002-840
Autoclavable Carts and Accessories, Metro™
Carts
Lab carts include two or three stainless steel autoclave shelves with aluminum split sleeves and four stainless steel swedged caster posts
Ordering information: Autoclavable casters must be ordered separately. Additional autoclave shelves and aluminum split sleeves are also ordered separately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty