Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boekel Orbital Shaker

Supplier: Boekel Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

270293 270292 270290 270200 270291
97004-394EA 236.12 USD InStock
97004-394 97004-392 97004-398 97004-396 97004-400
Boekel Orbital Shaker
Shakers and Mixers Orbital Shakers
The Boekel Scientific Orbital Shaker, with its compact low profile and small footprint, provides a smooth horizontal orbital motion for mixing in bottles, flasks, and beakers.

  • Variable speed: 50 to 300 rpm
  • Orbit: 18 mm
  • Timer with auto shut off: Set up to 99 hours or for continuous
  • Compact footprint (WxDxH): 10.5" x 11" x 4"
  • Interchangeable platforms to meet your needs

This unity can also be used in an incubator ranging from temperatures of 4°C to 40°C.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty