Would you like to log in again?
Finnpipette® Bottle Top Dispenser, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4421160 4421150 4421130 4421120 4421170 4421140
97004-406EA 983.71 USD InStock
97004-406 97004-416 97004-414 97004-408 97004-412 97004-410
Finnpipette® Bottle Top Dispenser, Thermo Scientific
Bottle Top Dispensers
Finnpipette Dispensers protect users even when handling aggressive liquids
  • Ergonomically designed
  • Non-slip piston handle
  • Excellent chemical and thermal resistance
  • Precise self-locking system in the volume setting
The dispensers are available in six models, covering volumes between 0.2 and 60mL. There is a universal fit for reagent bottles, and the dispensing head cap prevents dripping.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty