These ledges keep shelf contents orderly with 20.3 cm (8") high, pressure-fit dividers and prevent items from protruding or falling from shelves.
- Unique design allows them to attach to the top or underside of shelves
These are deisgned to be used with Super Erecta® shelving systems.
Ordering information: For designer color dividers, color must be specified at time of order. Designer color includes black matte, flame, hunter green, copper hammertone and silver hammertone. Additional clamping may be required to keep dividers in place. Contact your Avantor representative for more information.