Would you like to log in again?
Super Erecta Shelf® Utility Carts, MW Series, Metro™
Supplier: Metro International
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Super Erecta Shelf®
MW205 MW606 MW604 MW711 MW606 MW401 MW404 MW708 MW205 MW707 MW203 MW712 MW601 MW402 MW104 MW607 MW204 MW108 MW206 MW105 MW203 MW608 MW605 MW108 MW106 MW703 MW204 MW705 MW708 MW602 MW703 MW603 MW602 MW601 MW611 MW605 MW612 MW705 MW208 MW702 MW402 MW706 MW103 MW603 MW706 MW608 MW612 MW707 MW104 MW712 MW406 MW702 MW704 MW704 MW401 MW701 MW103 MW403 MW105 MW604 MW206 MW403 MW607 MW208 MW406 MW711 MW701 MW611 MW404 MW106
19766-566EA 600.42 USD InStock
CA22233-334 CA22233-338 CA22233-336 CA19766-014 19766-566 CA19766-574 97005-798 CA19766-572 CA97005-842 CA19766-570 CA97005-844 CA19766-578 CA19766-016 CA19766-576 19766-568 97005-790 97005-794 97005-796 97005-788 19766-554 CA97005-796 19766-552 CA97005-798 19766-550 CA19766-568 CA19766-566 22233-346 22233-344 22233-348 19766-580 CA97005-790 22233-342 97005-742 CA97005-794 22233-340 97005-854 CA97005-742 CA19766-552 CA97005-788 97005-856 CA19766-550 19766-584 19766-582 19766-548 CA19766-554 19766-546 22233-336 CA22233-344 22233-334 CA22233-342 CA22233-348 22233-338 CA22233-346 CA22233-340 19766-576 19766-016 97005-842 19766-574 CA19766-584 CA97005-854 97005-844 19766-014 CA97005-856 CA19766-582 19766-572 19766-570 CA19766-580 CA19766-548 CA19766-546 19766-578
Super Erecta Shelf® Utility Carts, MW Series, Metro™
Carts
These utility carts are designed for long life
All 45.7cm (18") wide carts have 10.1cm (4") light-duty casters. The 53.3cm (21") and 61cm (24") wide carts have 12.7cm (5") light-duty casters. All carts include 8.9cm (31/2") diameter donut bumpers. Overall height (to top of handle) is 101cm (40").
MW100 series carts feature two stainless steel flat shelves and stainless steel handles. MW200 series carts feature three stainless steel flat shelves and stainless steel handles. MW400 series carts feature two chrome wire shelves, one stainless steel flat shelf, and chrome handles. Solid shelf can be positioned on top, middle, or bottom. MW600 series carts feature two wire shelves in chrome or stainless steel with handles of matching material. MW700 series carts feature three wire shelves in chrome or stainless steel, with handles of matching material.
Flat shelves are ideal for transporting materials that require an expanse of flat, solid surface. They feature a raised edge, which permits fast cleanup of spilled liquids, and double-thick 18-gauge steel at the edges for rigidity and strength. Wire shelves have a bright, modern, sanitary appearance. Their open construction minimizes dust accumulation and maximizes visibility and air circulation.
They are maintenance-free and easy to maneuver in small areas.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty