Would you like to log in again?
Deck-Mounted Gooseneck Faucet with Foot-Operated Mixing Valve, WaterSaver Faucet
Supplier: WaterSaver Faucet
L181WSA-3001
97006-602EA 578.11 USD InStock
97006-602
Deck-Mounted Gooseneck Faucet with Foot-Operated Mixing Valve, WaterSaver Faucet
Faucets and Sinks Faucets
Deck-mounted, rigid 15.2 cm (6") gooseneck faucet comes with floor-mounted, foot-operated mixing valve for hot and cold water.
Faucet features a 9.5 mm (3/8") IPS female outlet, removable 10-serration hose end, 9.5 mm (3/8") female inlet, and forged brass base with polished chrome-plated finish. Mixing valve consists of self-closing renewable valve units with stainless steel valve stem, forged brass body with polished chrome-plated finish, cast brass pedals mounted on a hinged bracket, two 12.7 mm (1/2") NPT female inlets, and a 12.7 mm (1/2") NPT female outlet.
Certifications: CSA certified. Complies with ANSI/ASME A112.18.1M.
Ordering information: Faucet inlet hardware includes mounting shank, locknut, and washer. Mounting shank requires assembly and valve comes fully assembled.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty