Would you like to log in again?
CarePac® Test Weight Sets, METTLER TOLEDO®
Supplier: Mettler-Toledo
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CarePac®
11123045 11123147 30550613 11123151 11123011 11123103 11123107 11123056 11123110 11123157 11123108 11123111 11123009 11123054 30550616 11123149 11123059 11123011 11123153 11123151 11123150 11123145 11123047 11123104 11123158 11123008 11123107 11123010 11123052 11123156 11123057 11123008 11123048 11123045 11123146 11123155 11123055 11123044 11123106 11123110 11123052 11123105 30550614 11123058 11123153 11123156 11123159 11123152 11123049 11123145 11123146 11123148 11123101 11123051 11123053 11123056 11123053 11123054 11123101 11123144 11123048 11123050 11123108 11123102 11123157 11123049 11123044 11123111 11123057 11123109 11123154 11123055 11123150 11123046 11123100 11123147 11123148 11123100 11123144 11123010 11123059 11123102 11123051 11123009 11123058 11123155 11123149 11123159 11123047 11123109 11123152 11123154 11123004 11123050 11123046 30550649 11123158 11123103
97007-254EA 494 USD InStock
CA97007-256 CA97007-258 97007-254 CA97007-262 97007-256 CA97007-264 97007-250 97007-252 CA97007-260 97019-014 CA97007-308 97007-258 CA97007-266 CA97007-268 CA97007-274 97007-264 97007-300 CA97007-276 97007-266 CA97007-270 97007-260 CA97007-272 97007-262 97027-716 97027-718 CA97007-252 CA97007-254 CA97007-250 CA97027-718 CA97027-716 97007-328 CA97007-336 CA97007-338 97007-324 CA97007-332 97007-326 CA97007-334 97007-298 CA97007-340 97007-330 CA97007-342 97007-332 97007-294 97007-296 97007-290 97007-292 97007-338 CA97007-344 97007-334 97007-336 97007-342 97007-344 97007-340 CA97007-318 97007-306 CA97007-314 97007-308 CA97007-316 CA97007-278 CA97007-310 97007-268 97007-304 CA97007-312 97007-276 CA97007-284 97007-278 CA97007-320 CA97007-286 97007-310 97007-272 CA97007-280 97007-274 CA97007-282 97007-270 CA97007-326 97007-316 CA97007-328 97007-318 CA97007-322 CA97007-288 97007-312 CA97007-324 97007-314 97007-320 CA97007-296 97007-286 97007-322 CA97007-330 97007-288 CA97007-292 97007-282 CA97007-294 97007-284 CA97007-290 97007-280 97019-008 97019-006 97019-004
CarePac® Test Weight Sets, METTLER TOLEDO®
Weights Calibration Weights
These sets enable routine testing of balances by using two weights at maximum and minimum loads
  • Routine testing of balances
  • Multiple configurations
Weights are available in both ASTM and OIML configurations and are specified to validate process tolerances up to 0.03%. Sets include ergonomic tweezers and cleaning cloths. Customized third weights are available for customer-specific testing, such as minimum weight determination. Specially-designed cases house both types of weights and all equipment. Additional space is provided to accommodate a third weight.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty