Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benchtop Centrifuge, 5810

Supplier: Eppendorf
022628185 022625004 022628157 022627007 022628179 022625004 022628056 022627066 022628157 022628078 022628185 022628179 022628090 022628090 022627110 022628056 022627110 022628078
89210-986EA 16611.71 USD InStock
CA11017-364 CA11017-370 CA89210-984 89210-986 75856-496 CA89210-982 89210-984 97007-376 89305-182 89305-170 CA89210-986 97007-372 CA11017-358 89305-176 CA89210-980 89210-982 CA53513-808 53513-808
Benchtop Centrifuge, 5810
Centrifuges Benchtop Centrifuges
The eppendorf centrifuge 5810 is the perfect day-to-day centrifuge for laboratories with medium-throughput requirements. It has an extremely compact footprint yet still offers the real versatility of being able to take both tube and plate formats.

  • Centrifuge lid with soft-touch lid closure
  • Low access height of 29 cm for easy loading and unloading of samples
  • Quiet operation to improve work environment
  • Compact footprint saves valuable bench space
  • Automatic rotor recognition and imbalance detection for maximum operational safety
  • Aerosol-tight Eppendorf QuickLock® caps and lids for easy, one-handed operation
  • Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 to 750 ml
  • Plate rotors for centrifugation of all types of MTP, PCR, cell culture, or Deepwell Plates
  • Fixed-angle rotors for high-speed applications in tubes from 0.2 to 85 ml

Centrifuge 5810, with its renowned quality and reliability, offers the most cost-effective solution for medium to high-throughput applications requiring speeds of up to 20,913×g (14,000 rpm). This benchtop centrifuge has been designed to fit into every laboratory and make routine work more comfortable thanks to its quiet operation, soft-touch lid closure and low access height for easy loading and unloading.

Experience true versatility with a broad selection of rotors and adapters that facilitate mixed loading of tubes, bottles and plates. A new generation of rotors increases the capacity of Centrifuge 5810 to a maximum of 4×750 ml (or 28×50/56×15 ml). The swing-bucket rotor and adapters accommodate tubes and bottles from 0.2 to 750 ml, and the fixed-angle rotor 0.2 to 85 ml tubes. The plate rotor is ready to carry out centrifugation of all types of multiwell plates, such as PCR, cell culture, or deep-well plates.

Max. Capacity Plates: 16×MTP/4×DWP
Rotors Available: 18

Certifications: UL Listed, Certificate of Quality/Conformity (ISO 9001, ISO 13485, ISO 14001).

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty