Would you like to log in again?
OIML Class 4 Individual Precision Weights, ASTM, Mettler Toledo
Supplier: Mettler-Toledo
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
11123628 11123621 11123593 11123597 11123616 11123609 11123585 11123607 11123627 11123601 11123587 11123599 11123591 11123611 11123602 11123615 11123623 11123603 11123594 11123605 11123618 11123617 11123608 11123600 11123591 11123608 11123590 11123626 11123619 11123590 11123589 11123612 11123627 11123584 11123585 11123615 11123592 11123609 11123624 11123586 11123625 11123589 11123613 11123614 11123621 11123596 11123584 11123610 11123586 11123613 11123610 11123614 11123616 11123592 11123606 11123611 11123620 11123587 11123588 11123604 11123612 11123607 11123622 11123588 11123598 11123595 11123625 11123629
97007-526EA 1510 USD InStock
CA97007-538 97007-526 CA97007-534 97007-528 CA97007-536 97007-522 CA97007-498 97007-566 97007-488 CA97007-530 97007-524 97007-568 CA97007-532 97007-496 97007-498 CA97007-540 97007-530 97007-492 97007-494 97007-490 CA97007-502 CA97007-546 97007-536 97007-538 CA97007-542 97007-532 CA97007-500 CA97007-544 97007-534 97007-540 97007-542 97007-508 97007-504 75852-894 97007-548 CA97007-556 97007-506 75852-896 75852-890 97007-500 97007-544 97007-546 97007-502 75852-892 97007-550 97007-552 CA97007-528 97007-518 CA97007-568 97007-558 97007-514 97007-516 CA97007-564 97007-510 CA97007-486 97007-554 97007-556 97007-512 CA97007-488 97007-562 CA97007-494 97007-520 97007-564 CA97007-496 97007-486 CA97007-490 97007-560 CA97007-492
OIML Class 4 Individual Precision Weights, ASTM, Mettler Toledo
Weights Calibration Weights
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
  • 316 stainless steel
  • Low magnetic permeability
  • High corrosion resistance
  • High gloss polish
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty