Would you like to log in again?
Class 1 and UltraClass Cal-Paks with NVLAP Calibration Certificate, Troemner
Supplier: Troemner
30391788 30391791 30391782 30391799 30391784 30391783 30391795 30391798 30390006 30391793 30391786 30391789 30391794 30391792 30391790 30391785 30390007 30391787
97008-030EA 1874.09 USD InStock
97008-030 97008-002 97008-022 97008-010 97007-966 97007-998 97007-978 97007-962 97007-994 97007-986 97007-974 97007-990 97007-982 97007-970 97008-006 97008-026 97008-014 97008-018
Class 1 and UltraClass Cal-Paks with NVLAP Calibration Certificate, Troemner
Weights Calibration Weights
Troemner Cal-Paks consist of high quality precision mass standards with the lowest measurement uncertainties, packaged into a versatile weight case
Cal-Paks enable users to meet all regulatory and audit needs, including ISO, FDA, GMP, GLP, DOD, ANSI/NCSL Z540-1, 10CFR50, 21CFR, and USP 41. Each Cal-Pak contains three ASTM Alloy 8 Class 1 or UltraClass weights. The trio of weights include the manufacturer's recommended calibration weight, 10% of the calibration weight, and the recommended minimum weight determined by the balance's readability and published standard deviation under normal conditions as recommended by USP 41. These versatile and high precision weights can be used with all manufacturer's makes and models. Weights are supplied in a rugged polypropylene case, with each individual weight in its own high quality polycarbonate case. Forceps, brush, and glove are also provided. Manufactured in the United States.
Order Now
ORDER
SERVICES
Services: VWR Catalyst™ Services provides convenient and fast re-calibration services within 6 business days for your weights. Please contact us at 1.888.793.2300 or VWRCATALYST@vwr.com for a quote or more information. If your weights are found to be out of tolerance, we will make the proper adjustments to bring them back into tolerance. To prevent loss and ensure the timely processing and integrity of your precision weights and/or calibration devices, please do not pack in foam peanuts. Please avoid packing your items with foam, packing peanuts or other material that cannot be recycled.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty