Would you like to log in again?
Orion™ High Performance Ammonia Ion Selective Electrodes, Thermo Scientific
Supplier: Thermo Scientific
Orion™
9512HPBNWP 9512HPBNWP
97008-300EA 1318.63 USD InStock
CA97008-300 97008-300
Orion™ High Performance Ammonia Ion Selective Electrodes, Thermo Scientific
Electrodes ISE Electrodes
The high performance ammonia ion selective electrode (ISE) allows fast, simple, economical, and accurate measurements of dissolved ammonia in aqueous solutions.
  • Response time of 1 minute in sample concentrations of 1 ppm and higher; fast results make it easy to combine functionality with autosamplers
  • Keying feature on the electrode body prevents membrane from twisting or breaking during assembly
  • Choice of preassembled outer body with membrane cap or loose membranes
  • New wastewater applications developed using known addition and direct measurement techniques
  • EPA-compliant method
  • Waterproof BNC connector
This gas sensing electrode can also be used to measure the ammonium ion after conversion to ammonia or to measure organic nitrogen after Kjeldahl digestion of the sample. Sample colour and turbidity do not affect the measurement and samples do not need to be distilled. Almost all anions, cations and dissolved species, other than volatile amines, do not interfere.
Ordering information: Supplied with 1 m cable, 20 loose membranes, one preassembled outer body and membrane cap, two 60 ml bottles of electrode filling solution, spout cap, and tweezers.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty